Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113248.2 | NP_001106719.1 | 2 | 237 | ||
NM_009188.4 | NP_033214.2 | 2 | 237 | |||
GenBank mRNA | AF038848.1 | - | 2 | 217 | ||
AK146847.1 | - | 2 | 220 | |||
AK159076.1 | - | 2 | 220 | |||
AK165592.1 | - | 2 | 222 | |||
AK169433.1 | - | 2 | 222 | |||
AK173005.1 | - | 2 | 206 | |||
BC020049.1 | - | 2 | 201 | |||
BC021160.1 | - | 2 | 220 | |||
BC096367.1 | - | 2 | 205 | |||
BC129859.1 | - | 2 | 209 | |||
L38622.2 | - | 2 | 209 |