Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007967.2 | NP_031993.1 | 1 | 213 | ||
GenBank mRNA | BC061467.1 | - | 351 | 6827284 | ||
M93128.1 | - | 1 | 213 |