Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001631.4 | NP_001622.2 | 2 | 246 | ||
GenBank mRNA | AK313163.1 | - | 2 | 235 | ||
BC132678.1 | - | 2 | 188 | 40146700 | ||
DA376676.1 | - | 2 | 246 | |||
DA925041.1 | - | 2 | 235 | |||
M15694.1 | - | 2 | 158 | |||
M31008.1 | - | 1 | 248 |