siRNA Details
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007808.4 | NP_031834.1 | 3 | 350 | ||
GenBank mRNA | AK002732.1 | - | 3 | 352 | ||
AK003168.1 | - | 3 | 379 | |||
AK005336.1 | - | 3 | 422 | |||
AK010651.1 | - | 3 | 354 | |||
AK088098.1 | - | 3 | 351 | |||
AK143887.1 | - | 3 | 351 | |||
AK168017.1 | - | 3 | 351 | |||
BC034363.1 | - | 3 | 334 | 2650742 | ||
BC092213.1 | - | 2 | 299 | 6845432 | ||
BC094054.1 | - | 3 | 339 | 2865963 |

More Information
Other Species Targeted

