Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009400.2 | NP_033426.1 | 2 | 212 | ||
NM_021985.2 | NP_068820.1 | 2 | 212 | |||
GenBank mRNA | AF229432.1 | - | 2 | 212 | ||
AF229433.1 | - | 2 | 212 | |||
AF229434.1 | - | 2 | 212 | |||
AK020762.1 | - | 2 | 233 | |||
AK081878.1 | - | 2 | 201 | |||
AY157832.1 | - | 2 | 212 | |||
AY157833.1 | - | 2 | 212 | |||
U82534.1 | - | 2 | 212 |