Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008105.3 | NP_032131.2 | 1 | 702 | ||
GenBank mRNA | AB037597.1 | - | 1 | 93 | ||
AK054226.1 | - | 1 | 217 | |||
AK080061.1 | - | 1 | 268 | |||
AY236873.1 | - | 1 | 577 | |||
AY435149.1 | - | 1 | 93 | |||
BY068011.1 | - | 1 | 268 | |||
U68182.1 | - | 1 | 329 |