Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026641.2 | NP_080917.1 | 3 | 270 | ||
GenBank mRNA | AK042231.1 | - | 3 | 195 | ||
AK129339.1 | - | 3 | 198 | |||
AK147305.1 | - | 2 | 3797 | |||
BC034101.1 | - | 3 | 165 | 4501451 |