Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008105.3 | NP_032131.2 | 1 | 946 | ||
GenBank mRNA | AB037597.1 | - | 1 | 337 | ||
AK054226.1 | - | 1 | 461 | |||
AK080061.1 | - | 1 | 512 | |||
AY236873.1 | - | 1 | 821 | |||
AY435149.1 | - | 1 | 337 | |||
U68182.1 | - | 1 | 573 |