Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290789.1 | NP_001277718.1 | 2 | 856 | ||
NM_027917.3 | NP_082193.2 | 2 | 856 | |||
GenBank mRNA | AK004934.1 | - | 2 | 783 | ||
AK035462.1 | - | 2 | 817 | |||
AK085693.1 | - | 2 | 817 | |||
AK134332.1 | - | 2 | 784 | |||
BC005761.1 | - | 2 | 831 | 3601742 | ||
BC031598.1 | - | 2 | 748 | 3497170 |