Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077694.2 | NP_001071162.1 | 25 | 2977 | ||
NM_021469.3 | NP_067444.2 | 25 | 2795 | |||
GenBank mRNA | AF188290.2 | - | 24 | 2570 | ||
AK034587.1 | - | 4 | 588 | |||
AK131144.1 | - | 25 | 2792 |