Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290789.1 | NP_001277718.1 | 2 | 900 | ||
NM_027917.3 | NP_082193.2 | 2 | 900 | |||
GenBank mRNA | AK004934.1 | - | 2 | 827 | ||
AK035462.1 | - | 2 | 861 | |||
AK085693.1 | - | 2 | 861 | |||
AK134332.1 | - | 2 | 828 | |||
AK203999.1 | - | 1 | 27 | |||
BC005761.1 | - | 2 | 875 | 3601742 | ||
BC031598.1 | - | 2 | 792 | 3497170 |