Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290683.1 | NP_001277612.1 | 5 | 387 | ||
NM_028534.3 | NP_082810.1 | 5 | 657 | |||
GenBank mRNA | AK006817.1 | - | 477 | |||
AK014966.1 | - | 5 | 341 | |||
AK132868.1 | - | 5 | 384 | |||
BC006946.1 | - | 5 | 565 | 2810817 |