Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029303.2 | NP_083579.1 | 1 | 429 | ||
GenBank mRNA | AK005930.1 | - | 1 | 421 | ||
BC048629.1 | - | 1 | 429 | 6744051 |