Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177992.2 | NP_818773.1 | 2, 3 | 254 | ||
GenBank mRNA | AK017821.1 | - | 2, 3 | 549 | ||
AK031004.1 | - | 2, 3 | 256 | |||
AK075772.1 | - | 2, 3 | 212 | |||
BC003886.1 | - | 2, 3 | 213 | 3590411 | ||
BC024109.1 | - | 2, 3 | 210 | 5149353 | ||
BY099107.1 | - | 2, 3 | 255 |