Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290398.1 | NP_001277327.1 | 12, 13 | 1727 | ||
NM_133196.6 | NP_573459.1 | 12, 13 | 1805 | |||
GenBank mRNA | AF317552.1 | - | 12, 13 | 1773 | ||
AK032817.1 | - | 12, 13 | 1787 | |||
AK049971.1 | - | 12, 13 | 1718 | |||
AK088260.1 | - | 12, 13 | 1633 | |||
AK147438.1 | - | 10, 11 | 4799 | |||
EU616680.1 | - | 14, 15 | 1923 | |||
EU616681.1 | - | 12, 13 | 1695 | |||
EU616682.1 | - | 14, 15 | 1845 |