Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283028.1 | NP_001269957.1 | 5 | 775 | ||
NM_001283029.1 | NP_001269958.1 | 5 | 640 | |||
NM_001283030.1 | NP_001269959.1 | 4 | 499 | |||
NM_011462.3 | NP_035592.1 | 3 | 404 | |||
NM_146043.4 | NP_666155.1 | 5 | 700 | |||
GenBank mRNA | AK088127.1 | - | 5 | 641 | ||
AK134259.1 | - | 5 | 663 | |||
AK135816.1 | - | 3 | 399 | |||
AK135847.1 | - | 3 | 403 | |||
AK135933.1 | - | 3 | 402 | |||
AK136233.1 | - | 3 | 402 | |||
AK136251.1 | - | 3 | 403 | |||
AK139899.1 | - | 3 | 401 | |||
AK162171.1 | - | 3 | 401 | |||
AK162195.1 | - | 3 | 401 | |||
AK162258.1 | - | 3 | 398 | |||
AK163263.1 | - | 3 | 400 | |||
AK163293.1 | - | 3 | 401 | |||
AK163301.1 | - | 3 | 405 | |||
AK163308.1 | - | 3 | 403 | |||
AK163382.1 | - | 3 | 401 | |||
AK166528.1 | - | 3 | 404 | |||
AK166532.1 | - | 3 | 404 | |||
AK166580.1 | - | 3 | 363 | |||
AK166677.1 | - | 3 | 400 | |||
AK168544.1 | - | 5 | 701 | |||
BC016517.1 | - | 5 | 686 | 2654233 | ||
U48972.1 | - | 3 | 398 |