Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029586.2 | NP_083862.1 | 3, 4 | 458 | ||
NM_029606.3 | NP_083882.3 | 26, 27 | 3005 | |||
NM_145688.2 | NP_663721.2 | 5, 6 | 680 | |||
GenBank mRNA | AK005611.1 | - | 3, 4 | 459 | ||
AK006504.1 | - | 5, 6 | 666 | |||
BC005723.1 | - | 5, 6 | 492 | |||
BC048677.1 | - | 5, 6 | 652 | |||
BC086808.1 | - | 20, 21 | 2245 | 30632964 |