Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081030.2 | NP_001074499.2 | 39, 40 | 5696 | ||
NM_001170561.1 | NP_001164032.1 | 40, 41 | 5774 | |||
GenBank mRNA | AK076039.1 | - | 7, 8 | 941 | ||
AK129485.1 | - | 39, 40 | 5640 | |||
BC029156.1 | - | 10, 11 | 1332 | |||
BC098209.1 | - | 20, 21 | 3039 | |||
BC157935.1 | - | 39, 40 | 5586 | 9088115 | ||
BC158013.1 | - | 40, 41 | 5664 | 9007320 | ||
BC172094.1 | - | 39, 40 | 5586 | 9054380 |