Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033603.2 | NP_291081.2 | 12 | 1315 | ||
GenBank mRNA | AF320619.1 | - | 12 | 1315 | ||
AK002318.1 | - | 12 | 1310 | |||
AK017204.1 | - | 12 | 1330 | |||
AK088429.1 | - | 7 | 829 | |||
BC087954.1 | - | 12 | 1328 | 30119116 |