Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271353.1 | NP_001258282.1 | 12 | 1429 | ||
NM_144855.3 | NP_659104.1 | 12 | 1432 | |||
NM_178224.3 | NP_835742.1 | 12 | 1432 | |||
GenBank mRNA | AK077669.1 | - | 12 | 1393 | ||
AK085692.1 | - | 12 | 1398 | |||
BC013472.1 | - | 12 | 1358 | 4221477 | ||
BC013480.1 | - | 12 | 1368 | 4239757 | ||
BC022144.1 | - | 12 | 1393 | |||
BC026595.1 | - | 12 | 1365 | 4974777 |