Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173437.2 | NP_775613.2 | 23 | 4568 | ||
GenBank mRNA | AF307453.1 | - | 23 | 4928 | ||
AK036761.1 | - | 5 | 572 | |||
AK122458.1 | - | 23 | 4626 | |||
AK153742.1 | - | 6 | 889 | |||
BC080292.1 | - | 5 | 640 |