Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173788.3 | NP_776149.1 | 22 | 3343 | ||
GenBank mRNA | AK158195.1 | - | 17 | 1894 | ||
BC023420.1 | - | 9 | 984 | |||
BC037458.1 | - | 17 | 1799 | |||
BC042470.1 | - | 22 | 3343 | 5137261 | ||
BC049896.1 | - | 12 | 1438 |