Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174985.1 | NP_778150.1 | 1 | 895 | ||
GenBank mRNA | AB086171.1 | - | 1 | 895 | ||
AB089308.1 | - | 1 | 895 | |||
BC116914.1 | - | 1 | 945 | 40126233 | ||
BC119327.1 | - | 1 | 945 | 8734076 |
Set Membership: |
G-protein coupled receptor |