Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175036.4 | NP_778201.1 | 3 | 196 | ||
GenBank mRNA | AJ011565.1 | - | 3 | 169 | ||
AK009569.1 | - | 3 | 197 | |||
AK088848.1 | - | 1 | 13 | |||
AK152008.1 | - | 3 | 263 | |||
AK159489.1 | - | 3 | 262 | |||
AK160453.1 | - | 3 | 210 | |||
AK165821.1 | - | 3 | 255 | |||
AK168597.1 | - | 3 | 232 | |||
AK218321.1 | - | 1 | 44 | |||
BC004744.1 | - | 3 | 207 | 3583760 |