Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045959.1 | NP_001039424.1 | 2 | 264 | ||
NM_001045964.1 | NP_001039429.1 | 2 | 264 | |||
NM_016713.2 | NP_057922.2 | 2 | 264 | |||
NM_176893.2 | NP_795712.2 | 2 | 264 | |||
GenBank mRNA | AB035697.1 | - | 2 | 249 | ||
AB041925.1 | - | 2 | 250 | |||
BC052474.1 | - | 2 | 128 | 6833893 | ||
BC079549.1 | - | 2 | 122 | |||
CB245214.1 | - | 2 | 128 |