Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177324.2 | NP_796298.2 | 32 | 4399 | ||
GenBank mRNA | AK029568.1 | - | 3 | 402 | ||
BC015069.1 | - | 8 | 1115 |