Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159883.1 | NP_001153355.1 | 3 | 360 | ||
NM_001159884.1 | NP_001153356.1 | 3 | 308 | |||
NM_001159885.1 | NP_001153357.1 | 2 | 343 | |||
NM_020266.2 | NP_064662.2 | 3 | 360 | |||
NM_178055.4 | NP_835156.1 | 3 | 360 | |||
GenBank mRNA | AB028858.1 | - | 3 | 227 | ||
AK076061.1 | - | 3 | 296 | |||
AK156083.1 | - | 3 | 329 | |||
AK171494.1 | - | 3 | 306 | |||
BC011090.1 | - | 3 | 262 | 3968468 | ||
BC085162.1 | - | 2 | 580 | |||
BY092620.1 | - | 3 | 307 | |||
CJ174452.1 | - | 2 | 345 |