Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290625.1 | NP_001277554.1 | 4 | 564 | ||
NM_001290626.1 | NP_001277555.1 | 3 | 562 | |||
NM_177345.4 | NP_796319.1 | 4 | 890 | |||
GenBank mRNA | AK042154.1 | - | 3 | 552 | ||
AK052045.1 | - | 4 | 558 | |||
AK132263.1 | - | 4 | 554 | |||
AK148428.1 | - | 4 | 879 | |||
AK178100.1 | - | 2 | 138 | |||
BC043296.1 | - | 4 | 789 | |||
BC090644.1 | - | 3 | 464 | 30629263 | ||
BC096618.1 | - | 4 | 863 | 6392317 |