Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177396.1 | NP_796370.1 | 3 | 311 | ||
GenBank mRNA | AY184375.1 | - | 3 | 311 | ||
AY869696.1 | - | 3 | 311 |