Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mblac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177878.3 | NP_808546.1 | 1 | 820 | ||
GenBank mRNA | AK049381.1 | - | 1 | 821 | ||
BC038925.1 | - | 1 | 801 | 5374241 |