Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042591.1 | NP_001036056.1 | 1 | 330 | ||
GenBank mRNA | AK084951.1 | - | 1 | 341 | ||
AK088288.1 | - | 1 | 345 | |||
AK122499.1 | - | 1 | 317 | |||
BC044762.1 | - | 1 | 328 | |||
BC054844.1 | - | 1 | 330 | 6493441 | ||
BC098474.1 | - | 1 | 324 | 30094818 | ||
DQ861994.1 | - | 1 | 75 |