Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181407.4 | NP_852072.2 | 14 | 1814 | ||
GenBank mRNA | AK032196.1 | - | 14 | 1815 | ||
BC099935.1 | - | 14 | 1910 | 30097608 |