Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024186.5 | NP_077148.1 | 2 | 201 | ||
NM_024272.4 | NP_077234.2 | 2 | 201 | |||
GenBank mRNA | AK005150.1 | - | 2 | 136 | ||
AK010220.1 | - | 2 | 143 | |||
AK013177.1 | - | 2 | 127 | |||
AK039140.1 | - | 993 | ||||
AK051840.1 | - | 2 | 138 | |||
AK133699.1 | - | 2 | 145 | |||
AY037837.1 | - | 2 | 157 | |||
AY167988.1 | - | 2 | 111 | |||
BC002176.1 | - | 2 | 213 | 3486988 |