Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hba-x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010405.4 | NP_034535.1 | 2 | 288 | ||
GenBank mRNA | AK077590.1 | - | 2 | 287 | ||
BC051988.1 | - | 2 | 258 | 5711898 | ||
BY111754.1 | - | 2 | 289 | |||
M13125.1 | - | 1 | 96 | |||
M26898.1 | - | 2 | 246 |