Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026690.1 | NP_080966.1 | 4 | 449 | ||
GenBank mRNA | AK002587.1 | - | 4 | 449 | ||
BC019451.1 | - | 4 | 415 | 4194890 |