Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h2aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013549.2 | NP_038577.1 | 1 | 533 | ||
GenBank mRNA | AK002725.1 | - | 1 | 493 | ||
AK006728.1 | - | 1 | 492 | |||
BC053966.1 | - | 1 | 485 | |||
BC062255.1 | - | 1 | 476 | 1448126 | ||
BC064002.1 | - | 602 | ||||
BC089519.1 | - | 1 | 533 | 6771160 | ||
BC147778.1 | - | 1 | 530 | 9087498 | ||
BC147781.1 | - | 1 | 530 | 9087501 |
Gene Symbol: Hist2h2aa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178212.3 | NP_835584.1 | 1 | 573 |