Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195000.1 | NP_001181929.1 | 4 | 586 | ||
NM_001195035.1 | NP_001181964.1 | 4 | 586 | |||
NM_001310213.1 | NP_001297142.1 | 5 | 702 | |||
NM_001531.2 | NP_001522.1 | 4 | 586 | |||
XM_006711286.3 | XP_006711349.1 | 4 | 632 | |||
XM_011509463.2 | XP_011507765.1 | 3 | 358 | |||
GenBank mRNA | AF010446.1 | - | 3 | 430 | ||
AF010447.1 | - | 3 | 411 | |||
AF031469.1 | - | 3 | 413 | |||
AJ249778.1 | - | 3 | 430 | |||
AK222910.1 | - | 3 | 439 | |||
AK290374.1 | - | 3 | 450 | |||
AK304645.1 | - | 3 | 502 | |||
BX415555.2 | - | 4 | 538 | |||
U22963.1 | - | 3 | 430 |