Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001539.3 | NP_001530.1 | 3 | 381 | ||
GenBank mRNA | AK289623.1 | - | 3 | 309 | ||
AY186741.1 | - | 3 | 307 | |||
BC008182.1 | - | 3 | 294 | 2900892 | ||
BI462555.1 | - | 3 | 376 | |||
BM469197.1 | - | 3 | 282 | |||
BT007292.1 | - | 2 | 190 | |||
D13388.1 | - | 3 | 307 | |||
DA003255.1 | - | 3 | 383 | |||
L08069.1 | - | 3 | 272 |