Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007969.4 | NP_031995.3 | 3 | 358 | ||
GenBank mRNA | AK145032.1 | - | 3 | 254 | ||
BC010986.1 | - | 3 | 358 | 4038377 | ||
BE310436.1 | - | 3 | 219 | |||
EH099462.1 | - | 3 | 385 | |||
EH105860.1 | - | 3 | 385 | |||
EH109450.1 | - | 3 | 264 | |||
X93037.1 | - | 3 | 256 |