Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF354259.1 | - | 326 | |||
BC132126.1 | - | 4 | 364 | 40130403 | ||
BC132128.1 | - | 4 | 364 | 40130405 | ||
DQ237928.1 | - | 4 | 322 | |||
HQ713433.1 | - | 320 | ||||
M77677.1 | - | 4 | 365 | |||
X64719.1 | - | 293 |