Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039653.2 | NP_001034742.1 | 2 | 318 | ||
NM_010711.2 | NP_034841.2 | 2 | 235 | |||
GenBank mRNA | BC132554.1 | - | 2 | 232 | 40130831 | |
BC132556.1 | - | 2 | 232 | 40130833 | ||
BC150689.1 | - | 2 | 204 | 9087600 | ||
BY716979.1 | - | 2 | 299 | |||
L38248.1 | - | 2 | 284 | |||
L38857.1 | - | 2 | 235 |