Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019755.4 | NP_062729.1 | 1, 2 | 141 | ||
GenBank mRNA | AB031292.1 | - | 1, 2 | 159 | ||
AK003522.1 | - | 1, 2 | 196 | |||
AK011282.1 | - | 1, 2 | 195 | |||
AK012816.1 | - | 1, 2 | 236 | |||
AK154228.1 | - | 1, 2 | 205 | |||
BC083078.1 | - | 1, 2 | 184 | 6477560 | ||
BC092095.1 | - | 1, 2 | 160 | 3989985 | ||
BC119088.1 | - | 1, 2 | 141 | 8733837 | ||
BC119090.1 | - | 1, 2 | 141 | 8733839 |