Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019690.2 | NP_062664.2 | 4, 5 | 1178 | ||
GenBank mRNA | AF107847.1 | - | 4, 5 | 1182 | ||
BY718446.1 | - | 3, 4 | 234 |