Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025983.3 | NP_080259.1 | 2 | 197 | ||
GenBank mRNA | AK007752.1 | - | 2 | 186 | ||
AK007809.1 | - | 2 | 186 | |||
AK010661.2 | - | 2 | 190 | |||
AK012444.1 | - | 2 | 187 | |||
AK131585.1 | - | 660 | ||||
AK160387.1 | - | 2 | 184 | |||
AY341882.1 | - | 2 | 169 | |||
BC024339.1 | - | 2 | 189 | 4981796 | ||
BC046612.1 | - | 1 | 1480 | |||
CJ322223.1 | - | 2 | 197 | |||
W78532.1 | - | 2 | 172 |