Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111279.1 | NP_001104749.1 | 1, 2 | 214 | ||
NM_027057.3 | NP_081333.1 | 1, 2 | 214 | |||
GenBank mRNA | AK005937.1 | - | 1, 2 | 178 | ||
AK007215.1 | - | 1, 2 | 215 | |||
AK157204.1 | - | 1, 2 | 173 | |||
AK159710.1 | - | 1, 2 | 185 | |||
AK165554.1 | - | 1, 2 | 187 | |||
AK220391.1 | - | 1, 2 | 148 | |||
BC025226.1 | - | 1, 2 | 128 | 5011404 |