Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030742.1 | NP_109667.1 | 2 | 1273 | ||
GenBank mRNA | AF324866.1 | - | 2 | 1273 | ||
BC119575.2 | - | 1 | 1102 | 40058043 |
Gene Symbol: Vmn1r187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167568.1 | NP_001161040.1 | 1 | 388 |