Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001646.2 | NP_001637.1 | 3 | 420 | ||
GenBank mRNA | AV690709.1 | - | 3 | 369 | ||
BC020723.1 | - | 3 | 384 | 4700462 | ||
DB185657.1 | - | 3 | 420 |
Gene Symbol: APOC4-APOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037932.1 | 3 | 420 | |||
GenBank mRNA | AK126257.1 | - | 3 | 420 |