Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133962.3 | NP_598723.3 | 20 | 4620 | ||
GenBank mRNA | AK083608.1 | - | 1 | 397 | ||
AK172963.1 | - | 12 | 4597 | |||
BC009156.1 | - | 1 | 182 | |||
BC091746.1 | - | 1 | 182 | |||
BM935742.1 | - | 1 | 440 |