Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001669.3 | NP_001660.2 | 3 | 403 | ||
NM_009589.3 | NP_033667.2 | 3 | 403 | |||
XM_005274514.2 | XP_005274571.1 | 3 | 373 | |||
XM_005274515.2 | XP_005274572.1 | 3 | 373 | |||
GenBank mRNA | AF160499.1 | - | 3 | 303 | ||
AK314479.1 | - | 3 | 367 | |||
BC020229.1 | - | 331 | ||||
BC080179.1 | - | 3 | 306 | |||
DA869167.1 | - | 3 | 403 | |||
X83572.1 | - | 3 | 379 |