Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001688.4 | NP_001679.2 | 3 | 516 | ||
GenBank mRNA | AK223018.1 | - | 3 | 141 | ||
AK291077.1 | - | 3 | 173 | |||
AK308266.1 | - | 4 | 552 | |||
AK313541.1 | - | 3 | 143 | |||
AY871272.1 | - | 3 | 150 | |||
BC005366.1 | - | 3 | 161 | 3839745 | ||
BC005960.1 | - | 3 | 153 | 4049491 | ||
BC016350.1 | - | 3 | 192 | 4074170 | ||
X60221.1 | - | 3 | 127 |